DAPT

Lieferant: Thermo Fisher Scientific

Synonyme: N-[N-(3,5-Difluorophenacetyl-L-alanyl)]-S-phenylglycine t-Butyl Ester, tert-Butyl (S)-2-((S)-2-(2-(3,5-difluorophenyl)acetamido)propanamido)-2-phenylacetate, (3,5-Difluorophenylacetyl)-Ala-Phg-OtBu, (3,5-Difluorophenylacetyl)-L-alanyl-L-2-phenylglycine tert-Butyl Ester

J65864.MA
J65864.MAEA 190 EUR
J65864.MA
DAPT
DAPT

CAS No.: 208255-80-5 Molecular Formula: C23H26F2N2O4 Formula Weight: 432.50 Storage Temperature: -30°C to -10°C Physical Form: Lyophilized Sensitivity: Light Sensitive Appearance: White MDL No.: MFCD04974585


Applications: A cell-permeable dipeptide that suppresses A production by blocking -secretase

Formel: C₂₃H₂₆F₂N₂O₄
MDL: MFCD04974585
CAS-Nummer: 208255-80-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR