N-Acetyl-Asp-Glu-Val-Asp p-nitroanilide
Lieferant: Thermo Scientific
J65878.MB
J65878.MBEA
290
EUR
J65878.MB
N-Acetyl-Asp-Glu-Val-Asp p-nitroanilide
Proteine und Peptide
A colorimetric substrate for caspase-3.
CAS No.: 189950-66-1
Molecular Formula: C26H34N6O13
Formula Weight: 638.58
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White
MDL No.: MFCD00792707
Applications: A colorimetric substrate for caspase-3
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...