5'-O-(4,4'-Dimethoxytrityl)-2'-deoxyuridine 98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
1-((2R,4S,5R)-5-((Bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-4-hydroxytetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione
J65960.03
J65960.03EA
131
EUR
J65960.03
5'-O-(4,4'-Dimethoxytrityl)-2'-deoxyuridine 98%
5'-O-(4,4'-Dimethoxytrityl)-2'-deoxyuridine
Purity: 98%
CAS No.: 23669-79-6
Molecular Formula: C30H30N2O7
Formula Weight: 530.57
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White to off-white
Solubility: Soluble in acetonitrile at 20mg/ml
MDL No.: MFCD00063410
Formel:
C₃₀H₃₀N₂O₇ Molecular Weight: 530,57 g/mol |
MDL:
MFCD00063410 CAS-Nummer: 23669-79-6 EINECS: 245-814-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...