(6R,7R)-7-{[(2Z)-2-(2-Amino-1,3-thiazol-4-yl)-2-(methoxyimino)acetyl]amino}-3-(6,7-dihydro-5H-cyclopenta[b]pyridinium-1-ylmethyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-en-2-carboxylatsulfat 95%
Lieferant: Thermo Fisher Scientific
Purity: 95%
CAS No.: 98753-19-6
Molecular Formula: C22H22N6O5S2.H2SO4
Formula Weight: 612.66
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White to off-white
Melting Point: 198-202°
Solubility: Soluble in DMSO
MDL No.: MFCD11864966
Applications: A fourth generation cephalosporin antibiotic
Formel:
C₂₂H₂₄N₆O₉S₃ Schmelzpunkt: 198…202 °C |
MDL:
MFCD11864966 CAS-Nummer: 98753-19-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...