Cycloheximid 100 mg/ml in DMSO

Lieferant: Thermo Fisher Scientific
Gefahr


Filtriert über 0,2 µm-Filter

J66004.XF
J66004.XFEA 133 EUR
J66004.XF
Cycloheximid 100 mg/ml in DMSO
Cycloheximid

Filtered at 0.2 µm.


CAS No.: 66-81-9 Molecular Formula: C15H23NO4 Formula Weight: 281.35 Storage Temperature: 2°C to 8°C Flash Point: 87°0°(230°F) Physical Form: Liquid Appearance: Colourless to pale yellow UN No.: UN2810 MDL No.: MFCD00082346

Formel: C₁₅H₂₃NO₄
Flammpunkt: 87°0 °C (230 °F)
Lagertemperatur: Kühlschrank
MDL: MFCD00082346
CAS-Nummer: 66-81-9
EINECS: 200-636-0
UN: 2810
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3abdf7ad3f7b3ea9e67fba3b6f3970c6627794be&vid=6f07d0130a35bc66b22ecee29fc6c31b20823099&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR