Cefpodoxime proxetil 96%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: (6R,7R)-1-((Isopropoxycarbonyl)oxy)ethyl 7-((Z)-2-(2-aminothiazol-4-yl)-2-(methoxyimino)acetamido)-3-(methoxymethyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate

J66061.03
J66061.03EA 308 EUR
J66061.03
Cefpodoxime proxetil 96%
Cefpodoxime proxetil

Purity: 96% CAS No.: 87239-81-4 Molecular Formula: C21H27N5O9S2 Formula Weight: 557.60 Storage Temperature: 2°C to 8°C Physical Form: Powder Sensitivity: Light Sensitive Appearance: White to off-white Melting Point: 111-113° Solubility: Soluble in DMSO MDL No.: MFCD00865088


Applications: A broad spectrum antibiotic

Formel: C₂₁H₂₇N₅O₉S₂
Schmelzpunkt: 111…113 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00865088
CAS-Nummer: 87239-81-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR