Sisomicin sulphate Salt 90%

Lieferant: Thermo Fisher Scientific
Gefahr

J66063.03
J66063.03EA 220 EUR
J66063.03
Sisomicin sulphate Salt 90%
Sisomicin sulphate Salt

Sisomicin is a broad spectrum aminoglycoside antibiotic that targets the 30S ribosomal subunit resulting in an inability to read mRNA. Sisomicin is related to gentamicin, but is more effective against Pseudomonas aeruginosa.


  • White to off-white


Soluble in water


Bitte beachten For research use only. Not intended for animal or human diagnostic or therapeutic use.

Formel: (C₁₉H₃₇N₅O₇)₂·5H₂SO₄
Siedepunkt: 155 °C
Schmelzpunkt: 70…73 °C
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Kühlschrank
MDL: MFCD00058308
CAS-Nummer: 53179-09-2
EINECS: 258-414-4
Merck Index: 14,8550

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR