Ribostamycin sulphate 630 µg/mg

Lieferant: Thermo Fisher Scientific
Gefahr

J66155.MD J66155.03
J66155.MDEA 213 EUR
J66155.MD J66155.03
Ribostamycin sulphate 630 µg/mg
Ribostamycin sulphate

A broad spectrum antibiotic targeting both Gram-positive and Gram-negative bacteria.


  • White to pale yellow powder


Soluble in water.


Bitte beachten Research Use only: Not intended for animal or human diagnostic or therapeutic use.

Formel: C₁₇H₃₄N₄O₁₀·H₂SO₄
Schmelzpunkt: 175…180 °C
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Kühlschrank
MDL: MFCD00057496
CAS-Nummer: 53797-35-6
EINECS: 258-783-1
Merck Index: 14,8206

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR