Praziquantel 98%
Lieferant: Thermo Fisher Scientific
Achtung
|
Synonyme:
2-(Cyclohexylcarbonyl)-1,2,3,6,7,11b-hexahydro-4H-pyrazino[2,1-a]isoquinolin-4-one, 2-(Cyclohexanecarbonyl)-2,3,6,7-tetrahydro-1H-pyrazino[2,1-a]isoquinolin-4(11bH)-one
Purity: 98%
CAS No.: 55268-74-1
Molecular Formula: C19H24N2O2
Formula Weight: 312.41
Storage Temperature: 2°C to 8°C
Flash Point:
Applications: An anti-helminthic compound
Formel:
C₁₉H₂₄N₂O₂ Molecular Weight: 312.41 g/mol Siedepunkt: 1377 °C Schmelzpunkt: 136…138 °C Flammpunkt: >110 °C (230 °F) |
MDL:
MFCD00058531 CAS-Nummer: 55268-74-1 EINECS: 259-559-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...