Praziquantel 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-(Cyclohexylcarbonyl)-1,2,3,6,7,11b-hexahydro-4H-pyrazino[2,1-a]isoquinolin-4-one, 2-(Cyclohexanecarbonyl)-2,3,6,7-tetrahydro-1H-pyrazino[2,1-a]isoquinolin-4(11bH)-one

J66188.06 J66188.14
J66188.06EA 77.3 EUR
J66188.06 J66188.14
Praziquantel 98%
Praziquantel

Purity: 98% CAS No.: 55268-74-1 Molecular Formula: C19H24N2O2 Formula Weight: 312.41 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Physical Form: Powder Appearance: White to off-white Melting Point: 136-138° Boiling Point: 1377° Solubility: Freely soluble in ethanol or dichloromethane. Slightly soluble in water Beilstein Reference: 761557 MDL No.: MFCD00058531


Applications: An anti-helminthic compound

Formel: C₁₉H₂₄N₂O₂
Molecular Weight: 312.41 g/mol
Siedepunkt: 1377 °C
Schmelzpunkt: 136…138 °C
Flammpunkt: >110 °C (230 °F)
MDL: MFCD00058531
CAS-Nummer: 55268-74-1
EINECS: 259-559-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR