Cefotetan disodium

Lieferant: Thermo Scientific
Gefahr

J66206.MC
J66206.MCEA 114 EUR
J66206.MC
Cefotetan disodium
Cefotetan disodium salt

A broad spectrum antibiotic.


CAS No.: 74356-00-6 Molecular Formula: C17H15N7Na2O8S4 Formula Weight: 619.58 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White to off-white Boiling Point: 798° Solubility: Freely soluble in aqueous solution MDL No.: MFCD00941449


Applications: A broad spectrum antibiotic

Formel: C₁₇H₁₅N₇Na₂O₈S₄
Molecular Weight: 619,568 g/mol
Siedepunkt: 798 °C
MDL: MFCD00941449
CAS-Nummer: 74356-00-6
EINECS: 277-834-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR