Roxithromycin 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: (3R,4S,5S,6R,7R,9R,11S,12R,13S,14R,E)-6-(((2S,3R,4S,6R)-4-(Dimethylamino)-3-hydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-14-ethyl-7,12,13-trihydroxy-4-(((2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyltetrahydro-2H-pyran-2-yl)oxy)-10-(((2-methoxyethoxy)methoxy)imino)-3,5,7,9,11,13-hexamethyloxacyclotetradecan-2-one

J66213.09
J66213.09EA 123 EUR
J66213.09
Roxithromycin 97%
Roxithromycin

Roxithromycin is used to block bacterial protein synthesis


  • White powder


Clinically, used to treat respiratory tract, urinary and soft tissue infections.


Soluble in water to 18µg/ml

Formel: C₄₁H₇₆N₂O₁₅
Molecular Weight: 837,04 g/mol
Schmelzpunkt: 111…118 °C
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Kühlschrank
MDL: MFCD00214389
CAS-Nummer: 80214-83-1
Merck Index: 14,8276

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR