Cefpodoxime 98%
Lieferant: Thermo Scientific
|
Gefahr
|
J66225.03
J66225.03EA
314
EUR
J66225.03
Cefpodoxime 98%
Cefpodoxime
Purity: 98%
CAS No.: 80210-62-4
Molecular Formula: C15H17N5O6S2
Formula Weight: 427.45
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Sensitivity: Light Sensitive
Appearance: Brown
Melting Point: 200-202°
Solubility: Soluble in DMSO
MDL No.: MFCD00864906
Formel:
C₁₅H₁₇N₅O₆S₂ Schmelzpunkt: 200…202 °C |
MDL:
MFCD00864906 CAS-Nummer: 80210-62-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...