Sulbactam 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Sulbactam acid, (2S,5R)-3,3-Dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic Acid 4,4-Dioxide

J66235.03 J66235.06
J66235.03EA 78 EUR
J66235.03 J66235.06
Sulbactam 98%
Sulbactam

Purity: 98% CAS No.: 68373-14-8 Molecular Formula: C8H11NO5S Formula Weight: 233.24 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White to pale brown Melting Point: 154-157° Solubility: Soluble in water at 18mg/ml MDL No.: MFCD00867005


Applications: Commonly used beta-lactam antibiotics to prevent degradation by beta-lactamase enzymes

Formel: C₈H₁₁NO₅S
Molecular Weight: 233.24 g/mol
Schmelzpunkt: 154…157 °C
MDL: MFCD00867005
CAS-Nummer: 68373-14-8
EINECS: 269-878-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR