Sulbactam 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Sulbactam acid, (2S,5R)-3,3-Dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic Acid 4,4-Dioxide
J66235.03
J66235.06
J66235.03EA
78
EUR
J66235.03
J66235.06
Sulbactam 98%
Sulbactam
Purity: 98%
CAS No.: 68373-14-8
Molecular Formula: C8H11NO5S
Formula Weight: 233.24
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White to pale brown
Melting Point: 154-157°
Solubility: Soluble in water at 18mg/ml
MDL No.: MFCD00867005
Applications: Commonly used beta-lactam antibiotics to prevent degradation by beta-lactamase enzymes
Formel:
C₈H₁₁NO₅S Molecular Weight: 233.24 g/mol Schmelzpunkt: 154…157 °C |
MDL:
MFCD00867005 CAS-Nummer: 68373-14-8 EINECS: 269-878-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...