Pefloxacine methanesulphonate dihydrate 95%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
1-Ethyl-6-fluoro-7-(4-methylpiperazin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid mesylate dihydrate, Pefloxacin mesylate dihydrate
J66285.09
J66285.09EA
47.4
EUR
J66285.09
Pefloxacine methanesulphonate dihydrate 95%
Pefloxacine methanesulphonate dihydrate
A broad spectrum antibiotic that targets Gram-negative and -positive bacteria, and also some mycoplasma species
- Light sensitive
Off-white powder
Freely soluble in water
Formel:
C₁₇H₂₀FN₃O₃·CH₄O₃S·2H₂O Molecular Weight: 333,4 · 96,1 · 36.0 g/mol Siedepunkt: 529 °C Schmelzpunkt: 271 °C Flammpunkt: >110 °C (230 °F) Lagertemperatur: Kühlschrank |
MDL:
MFCD01685696 CAS-Nummer: 149676-40-4 EINECS: 274-613-9 Merck Index: 14,7066 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...