N-(5-Aminopentyl)biotinamide trifluoroacetate salt

Lieferant: Thermo Scientific

Synonyme: Biotin-cadaverine

J66382.MF
J66382.MFEA 233 EUR
J66382.MF
N-(5-Aminopentyl)biotinamide trifluoroacetate salt
N-(5-Aminopentyl)biotinamide trifluoroacetate salt

Molecular Formula: C17H29F3N4O4S Formula Weight: 442.50 Physical Form: Solid Sensitivity: Moisture and Light Sensitive Appearance: White to off-white MDL No.: MFCD03452814

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR