Meticillin sodium salt ≥99%

Lieferant: Thermo Fisher Scientific
Gefahr

J66414.03
J66414.03EA 262 EUR
J66414.03
Meticillin sodium salt ≥99%
Meticillin sodium salt

Purity: 90% CAS No.: 132-92-3 Molecular Formula: C17H19N2NaO6S Formula Weight: 402.40 Storage Temperature: -30°C to -10°C Flash Point: 110°(230°F) Physical Form: Powder Sensitivity: Light Sensitive Appearance: White to off-white Melting Point: 196-197° dec. Boiling Point: 640° Solubility: Slightly soluble in acetone. Soluble in ethanol or water MDL No.: MFCD07787409


Applications: Targets primarily the cell wall of gram positive organisms especially Staphylococcus aureus

Formel: C₁₇H₁₉N₂NaO₆S
Siedepunkt: 640 °C
Flammpunkt: >110 °C (230 °F)
MDL: MFCD07787409
CAS-Nummer: 132-92-3
EINECS: 205-083-9
Merck Index: 14,5969

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR