Meropenem Trihydrat 98-101%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
(4R,5S,6S)-3-(((3S,5S)-5-(Dimethylcarbamoyl)pyrrolidin-3-yl)thio)-6-((R)-1-hydroxyethyl)-4-methyl-7-oxo-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid trihydrate
A broad spectrum antibiotic targeting a wide range of bacteria.
Purity: 98-101%
CAS No.: 119478-56-7
Molecular Formula: C17H25N3O5S
Formula Weight: 383.46
Storage Temperature: 2°C to 8°C
Flash Point:
Off-white to pale yellow powder
Soluble in aqueous solution at approximately 5mg/ml
Formel:
C₁₇H₃₁N₃O₈S Molecular Weight: 437,51 g/mol Siedepunkt: 627 °C Flammpunkt: >110 °C (230 °F) Lagertemperatur: Kühlschrank |
MDL:
MFCD08600005 CAS-Nummer: 119478-56-7 Merck Index: 14,5900 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...