Cefotiam dihydrochloride

Lieferant: Thermo Scientific
Achtung

Synonyme: (6R,7R)-7-(2-(2-Aminothiazol-4-yl)acetamido)-3-(((1-(2-(dimethylamino)ethyl)-1H-tetrazol-5-yl)thio)methyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid dihydrochloride, Cefotiam HCl

J66529.MC
J66529.MCEA 136 EUR
J66529.MC
Cefotiam dihydrochloride
Cefotiam dihydrochloride

A broad spectrum antibiotic


CAS No.: 66309-69-1 Molecular Formula: C18H23N9O4S3.HCl Formula Weight: 562.09 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Physical Form: Powder Appearance: White Boiling Point: 94° Solubility: Soluble in water MDL No.: MFCD09878605


Applications: A broad spectrum antibiotic

Formel: C₁₈H₂₄ClN₉O₄S₃
Molecular Weight: 562,079 g/mol
Siedepunkt: 94 °C
Flammpunkt: >110 °C (230 °F)
MDL: MFCD09878605
CAS-Nummer: 66309-69-1
EINECS: 266-312-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR