Moxifloxacin hydrochloride 97%

Lieferant: Thermo Fisher Scientific

Synonyme: 1-Cyclopropyl-6-fluoro-7-((4aS,7aS)-hexahydro-1H-pyrrolo[3,4-b]pyridin-6(2H)-yl)-8-methoxy-4-oxo-1,4-dihydroquinoline-3-carboxylic acid hydrochloride, Moxifloxacin HCl

J66626.MC
J66626.MCEA 216 EUR
J66626.MC
Moxifloxacin hydrochloride 97%
Moxifloxacin hydrochloride

Purity: 97% CAS No.: 186826-86-8 Molecular Formula: C21H24FN3O4.HCl Formula Weight: 437.89 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Physical Form: Powder Appearance: Yellow Odour: Odourless Boiling Point: 636° Solubility: Sparingly soluble in water. Soluble in DMSO Beilstein Reference: 8377447 MDL No.: MFCD00949117


Applications: A broad spectrum antibiotic commonly used against bacteria which cause sinus and lung infections

Formel: C₂₁H₂₅ClFN₃O₄
Molecular Weight: 437.896 g/mol
Siedepunkt: 636 °C
Flammpunkt: >110 °C (230 °F)
MDL: MFCD00949117
CAS-Nummer: 186826-86-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR