Bisacrylamid 99+% für die Elektrophorese

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: N,N'-Methylendiacrylamid, BIS

J66710.14 J66710.22 J66710.30
J66710.14EA 65.9 EUR
J66710.14 J66710.22 J66710.30
Bisacrylamid 99+% für die Elektrophorese
Bisacrylamid

Purity: 99+% CAS No.: 110-26-9 Molecular Formula: C7H10N2O2 Formula Weight: 154.17 Physical Form: Powder Density: 1.235 Sensitivity: Air and Light Sensitive Appearance: White Melting Point: 300° Solubility: Soluble in water or methanol Beilstein Reference: 1706297 MDL No.: MFCD00008625

Formel: (H₂C=CHCONH)₂CH₂
Schmelzpunkt: >300 °C
Dichte: 1,235
Lagertemperatur: Raumtemperatur
MDL: MFCD00008625
CAS-Nummer: 110-26-9
EINECS: 203-750-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c1b72b0c3cd5bb916c65f9e2337c5c3b05ea1c91&vid=368f6bf0bc8e0b35ffc7a57d85f5dd210ff0b409&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR