Azithromycin dihydrate
Lieferant: Thermo Scientific
Synonyme:
Azithromycin hydrate, (2R,3S,4R,5R,8R,10R,11R,12S,13S,14R)-11-(((2S,3R,4S,6R)-4-(dimethylamino)-3-hydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-2-ethyl-3,4,10-trihydroxy-13-(((2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyltetrahydro-2H-pyran-2-yl)oxy)-3,5,6,8,10,12,14-heptamethyl-1-oxa-6-azacyclopentadecan-15-one dihydrate
A broad spectrum macrolide antibiotic structurally related to erythromycin. Azithromycin dihydrate has anti-immunomodulatory/anti-inflammatory properties, which make it useful in treating cystic fibrosis
- Air Sensitive
- Optical rotation: -47° (c=2 in ethanol)
It shows an azalide antimicrobial agent active in vitro against various pathogens.
White powder
Soluble in DMSO at 20mg/ml. Also soluble in chloroform or ethylene chloride
Formel:
C₃₈H₇₂N₂O₁₂·2H₂O Molecular Weight: 749,00 g/mol Siedepunkt: 717 °C Schmelzpunkt: 126 °C Flammpunkt: >110 °C (230 °F) Lagertemperatur: Kühlschrank |
MDL:
MFCD01862248 CAS-Nummer: 117772-70-0 |
Spezifikation Testergebnisse
Appearance (Color) | White to off-white |
Form | Powder |
Assay (unspecified) | 945 µg/mg min. |
Water Content (Karl Fischer Titration) | 4.0 - 5.0% |
Optical Rotation | -45° to -49° |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...