Mecillinam 90+%

Lieferant: Thermo Scientific

Synonyme: (2S,5R,6R)-6-(((E)-Azepan-1-ylmethylene)amino)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid

J66774.MC
J66774.MCEA 167 EUR
J66774.MC
Mecillinam 90+%
Mecillinam

Effective against gram-negative bacteria.


Soluble in water at approximately 1 mg/ml


Bitte beachten Research Use only: Not intended for animal or human diagnostic or therapeutic use.

Formel: C₁₅H₂₃N₃O₃S
Molecular Weight: 325,427 g/mol
Siedepunkt: 551 °C
Schmelzpunkt: 156 °C
Flammpunkt: >110 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00056869
CAS-Nummer: 32887-01-7
EINECS: 251-277-1
Merck Index: 14,00389

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR