Cefoxitin sodium 92,7-97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Mefoxin, Cefoxitin sodium salt, Sodium (6R,7S)-3-((carbamoyloxy)methyl)-7-methoxy-8-oxo-7-(2-(thiophen-2-yl)acetamido)-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate

J66800.03
J66800.03EA 182 EUR
J66800.03
Cefoxitin sodium 92,7-97%
Cefoxitin sodium

Purity: 92.7-97% CAS No.: 33564-30-6 Molecular Formula: C16H16N3NaO7S2 Formula Weight: 449.43 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Physical Form: Powder Appearance: White to off-white Melting Point: 160° Boiling Point: 843° Solubility: Soluble in water or methanol Optical Rotation: +210° (c=1 in methanol) MDL No.: MFCD00079042


Applications: A broad spectrum antibiotic

Formel: C₁₆H₁₆N₃NaO₇S₂
Molecular Weight: 449.428 g/mol
Siedepunkt: 843 °C
Flammpunkt: >110 °C (230 °F)
MDL: MFCD00079042
CAS-Nummer: 33564-30-6
EINECS: 251-574-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR