Pyrazinamid 98%
Lieferant: Thermo Fisher Scientific
Purity: 98%
CAS No.: 98-96-4
Molecular Formula: C5H5N3O
Formula Weight: 123.11
Storage Temperature: 2°C to 8°C
Flash Point:
Formel:
C₅H₅N₃O Schmelzpunkt: 189…191 °C Flammpunkt: >110 °C (230 °F) |
MDL:
MFCD00006132 CAS-Nummer: 98-96-4 EINECS: 202-717-6 Merck Index: 14,7956 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...