Coelenterazine (natürlich) , Aequorea sp.
Lieferant: Thermo Scientific
Synonyme:
CLZN
J66823.MA
J66823.MAEA
582
EUR
J66823.MA
Coelenterazine (natürlich) , Aequorea sp.
Coelenterazine
A sensitive and specific chemiluminescence probe of the superoxide anion.
CAS No.: 55779-48-1
Molecular Formula: C26H21N3O3
Formula Weight: 423.46
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Sensitivity: Moisture and Light Sensitive
Appearance: Yellow to dark orange
Solubility: Soluble in methanol or ethanol
MDL No.: MFCD00467176
Applications: A sensitive and specific chemiluminescence probe of the superoxide anion
Formel:
C₂₆H₂₁N₃O₃ Molecular Weight: 423,472 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00467176 CAS-Nummer: 55779-48-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...