Methyl-β-cyclodextrin (mixture of several Methylated)
Lieferant: Thermo Fisher Scientific
Methyl-β-cyclodextrin is used to remove cholesterol from cultured cells and to remove lipid rafts. Ideal for use in tissue and cell culture applications.
- Incompatible with heat, oxidising agents
The cavity diameter of β-cyclodextrins or β-glucopyranose unit compounds is well-suited for use with molecules the size of hormones, vitamins and many compounds frequently used in tissue and cell culture applications. For this reason, β-cyclodextrin is most commonly used as a complexing agent. It is used to increase the solubility of non-polar substances such as fatty acids, lipids, vitamins and cholesterol for use in cell culture applications.
White to off-white
Melting Point: 180-182°
Optical Rotation: +157.8° (c=1.5 in water)
Solubility: Soluble in water
Schmelzpunkt:
180…182 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00074980 CAS-Nummer: 128446-36-6 EINECS: 411-120-1 |
Spezifikation Testergebnisse
Appearance (Color) | White to off-white |
Form | Powder |
Optical Rotation | +150,0 ° to +165,5 °, c-1,5 in water |
Solubility | Soluble in water |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...