Mupirocin lithium ≥95%
Lieferant: Thermo Scientific
Synonyme:
Lithium 9-(((E)-4-((2S,3R,4R,5S)-3,4-dihydroxy-5-(((2S,3S)-3-((2S,3S)-3-hydroxybutan-2-yl)oxiran-2-yl)methyl)tetrahydro-2H-pyran-2-yl)-3-methylbut-2-enoyl)oxy)nonanoate
J66890.MC
J66890.MCEA
517
EUR
J66890.MC
Mupirocin lithium ≥95%
Mupirocin lithium
Purity: 95%
CAS No.: 73346-79-9
Molecular Formula: C26H43LiO9
Formula Weight: 506.55
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
Solubility: Soluble in water at approximately 10mg/ml
MDL No.: MFCD11977847
Applications: Commonly used to target gram positive bacteria
Formel:
C₂₆H₄₃LiO₉ Molecular Weight: 506,561 g/mol |
MDL:
MFCD11977847 CAS-Nummer: 73346-79-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...