Cefoxitin 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
J66891.03
J66891.03EA
170
EUR
J66891.03
Cefoxitin 98%
Cefoxitin
Purity: 98%
CAS No.: 35607-66-0
Molecular Formula: C16H17N3O7S2
Formula Weight: 427.45
Storage Temperature: 2°C to 8°C
Flash Point:
Applications: A broad spectrum antibiotic
Formel:
C₁₆H₁₇N₃O₇S₂ Molecular Weight: 427.446 g/mol Siedepunkt: 843 °C Schmelzpunkt: 149…150 °C Flammpunkt: >110 °C (230 °F) |
MDL:
MFCD00072014 CAS-Nummer: 35607-66-0 EINECS: 252-641-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...