Cycloheximid 95%, ultrarein
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
J66901.03
J66901.03EA
88
EUR
J66901.03
Cycloheximid 95%, ultrarein
Cycloheximid
Applications: A protein synthesis inhibitor in eukaryotic cells.
Applications: A protein synthesis inhibitor in eukaryotic cells
Formel:
C₁₅H₂₃NO₄ Siedepunkt: 492 °C Schmelzpunkt: 111…116 °C Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00082346 CAS-Nummer: 66-81-9 EINECS: 200-636-0 UN: 2811 ADR: 6.1,I Merck Index: 14,2728 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...