1,3,4,6-Tetra-O-acetyl-β-D-glucosamine hydrochloride

Lieferant: Thermo Fisher Scientific

Synonyme: 1,3,4,6-Tetra-O-acetyl-2-amino-2-deoxy-beta-D-glucopyranose hydrochloride, (2S,3R,4R,5S,6R)-6-(Acetoxymethyl)-3-aminotetrahydro-2H-pyran-2,4,5-triyl triacetate hydrochloride

J66975.18
J66975.18EA 841 EUR
J66975.18
1,3,4,6-Tetra-O-acetyl-β-D-glucosamine hydrochloride
1,3,4,6-Tetra-O-acetyl-beta-D-glucosamine hydrochloride

CAS No.: 10034-20-5 Molecular Formula: C14H21NO9.HCl Formula Weight: 383.78 Physical Form: Powder Appearance: White to pale brown Solubility: Soluble in water at 10mg/ml MDL No.: MFCD01075204

Formel: C₁₄H₂₂ClNO₉
Molecular Weight: 383.778 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01075204
CAS-Nummer: 10034-20-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR