Hyaluronic acid sodium salt (aus Streptococcus equi) 91%
Lieferant: Thermo Fisher Scientific
Synonyme:
Sodium hyaluronate, Hyaluronic acid sodium, HA sodium salt
J66993.03
J66993.03EA
88.7
EUR
J66993.03
Hyaluronic acid sodium salt (aus Streptococcus equi) 91%
Hyaluronic acid sodium salt
Bacterial hyaluronic acid (HA) is a natural non-sulfated high molecular weight glycosaminoglycan that may be used to study formulations of non-animal gels potentially useful for procedures such as soft tissue augmentation.
Molecular weight is greater than or equal to 1MDa
Applications: An anionic, nonsulfated glycosaminoglycan distributed widely throughout connective, epithelial, and neural tissues
Formel:
(C₁₄H₂₁NaNO₁₁)n Lagertemperatur: Gefrierschrank |
MDL:
MFCD00875848 CAS-Nummer: 9067-32-7 Merck Index: 14,4757 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...