Gold 99%, nanoparticles 150 nm reactant-free
Lieferant: Thermo Fisher Scientific
Synonyme:
Aurum
J67040.AC
J67040.ACEA
128
EUR
J67040.AC
Gold 99%, nanoparticles 150 nm reactant-free
Gold
Purity: 99%
CAS No.: 7440-57-5
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Dark brown to pink
MDL No.: MFCD00003436
Formel:
Au Molecular Weight: 196.97 g/mol Siedepunkt: 2800 Schmelzpunkt: 1063 Lagertemperatur: Kühlschrank |
MDL:
MFCD00003436 CAS-Nummer: 7440-57-5 Merck Index: 12,04526 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...