DNA molecular weight marker, wide range
Lieferant: Thermo Fisher Scientific
J67051.XI
J67051.XIEA 242 EUR
J67051.XI
DNA molecular weight marker, wide range
Elektrophorese-Reagenzien Molekulargewichtsmarker
This wide range molecular weight marker contains 60 ng/μl of 21 fragments from 100 bp to 12000 bp. The bands appear in 100 bp increments from 100 to 1000 bp and in 1000 bp increments from 1 to 12 kb. Bands at 1 kb and 5 kb stain brighter to aid identification.