Gold sodium citrate_0.1mg/ml+stab_supplied Stabilised, nanoparticles 15 nm OD1 (Abs. 520 nm)

Lieferant: Thermo Fisher Scientific

Synonyme: Aurum

J67307.AC
J67307.ACEA 91.7 EUR
J67307.AC
Gold sodium citrate_0.1mg/ml+stab_supplied Stabilised, nanoparticles 15 nm OD1 (Abs. 520 nm)
Gold

CAS No.: 7440-57-5 Storage Temperature: 2°C to 8°C Physical Form: Liquid Appearance: Pink to red MDL No.: MFCD00003436


Applications: For use in adsorption of thiolated ligands such as oligonucleotides and polyethylene glycols. Also, due to the increased stability, these gold nanoparticles are ideal for use in physical and optical applications.

Formel: Au
Molecular Weight: 196.97 g/mol
Siedepunkt: 2800
Schmelzpunkt: 1063
Lagertemperatur: Kühlschrank
MDL: MFCD00003436
CAS-Nummer: 7440-57-5
Merck Index: 12,04526

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR