Gold 95%, nanoparticles 90 nm OD1 (Abs. 564 nm)

Lieferant: Thermo Fisher Scientific

Synonyme: Aurum

J67426.AC
J67426.ACEA 87.5 EUR
J67426.AC
Gold 95%, nanoparticles 90 nm OD1 (Abs. 564 nm)
Gold

Purity: 95% CAS No.: 7440-57-5 Storage Temperature: 2°C to 8°C Physical Form: Liquid Appearance: Red to pink MDL No.: MFCD00003436


Applications: For development of peptide and protein gold conjugates for use in applications such as blotting, microscopy and TEM

Formel: Au
Molecular Weight: 196.97 g/mol
Siedepunkt: 2800
Schmelzpunkt: 1063
Lagertemperatur: Kühlschrank
MDL: MFCD00003436
CAS-Nummer: 7440-57-5
Merck Index: 12,04526

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR