YPD Bouillon für die Biotechnologie
Lieferant: VWR Chemicals
Genutzt für die Propagation und Versorgung von Hefen.
YPD broth is supplied as a pre-mixed powder which, when dissolved at 50 g/l of water, yields peptone (20 g/l), yeast extract (10 g/l), glucose (20 g/l).
Final pH of solution (25 °C): 6,5 ±0.5
Autoclave or sterile filter.