Microtube tube racks, foam
Lieferant: JAECE INDUSTRIES
FTR100
JAECFTR100EA 202 EUR
JAECFTR100
Microtube tube racks, foam
Gestelle Reagenzglasgestelle
Economical, charcoal gray, open-cell foam rack have a capacity for 50 tubes. Ideal for storage on benchtops, in incubators and refigerators/freezers. Can be used to protect valuable, fragile tubes during mailing/shipping. Ideal alternative to expensive plastic racks.
  • Racks will float for use in waterbaths
  • Stackable