Cole-Parmer® WB-400 Series Digital General-Purpose Water Bath
Lieferant: Cole-Parmer
12105-87
JENW12105-87EA 1690 EUR
JENW12105-87
Cole-Parmer® WB-400 Series Digital General-Purpose Water Bath
Bäder Wasserbäder, ohne Umwälzung
Get digital accuracy at an analogue price.
  • Recall settings at the touch of a button - program up to 5 commonly used set points
  • Know when process is complete with built-in timer
  • Easy-to-read TFT display shows choice of °C or °F
  • Features high-temperature alarm and safety shutdown
Cole-Parmer utility baths combine precise temperature control and excellent temperature stability to provide reliable heating. These baths feature dual thermostats - a primary microprocessor-based temperature controller that provides ±0,1 °C stability (±0,2 °C uniformity), and a secondary over-temperature safety sensor prevents samples from overheating. For optimal temperature accuracy, the primary controller can be calibrated to your reference thermometer.
The stainless steel tank is seamless - no welds to leak or rust - and the rounded corners provide an easy to clean interior. The high-dome, hinged polycarbonate lid tilts out of the way, while allowing complete view of the sample and condensation to drain back.
Lieferumfang: Supplied with domed lid and power cord.