Conductivity/TDS/°C meters, bench, 4510 / 4520
Lieferant: Cole-Parmer
451001 452001 35440-06
664-0112EA 913 EUR
664-0112 664-0113 JENW35440-06
Conductivity/TDS/°C meters, bench, 4510 / 4520
Elektrochemische Messgeräte Leitfähigkeitsmessgeräte
4510 is an easy-to-use meter offering the flexibility to meet the broadest range of applications and for those where greater accuracy is required. With automatic conductivity standard recognition and endpoint detection. 4520 is a high specification laboratory conductivity/resistivity/TDS/salinity/°C meter that offers additional accuracy with 1, 2 or 3 point conductivity calibration across a wide measurement range. The dedicated 'pure water' mode ensures the optimum accuracy for this difficult application.
  • Simultaneous display of conductivity or TDS and temperature
  • Automatic selection of range for best resolution
  • Calibration by cell constant or standard solutions
  • Storage of up to 32 results (4510) or 500 results (4520)
  • 4510: Bi-directional RS232 link to printer or PC
  • 4520: Infra-red data link (IrDA) for connection to printer and RS232 link to connect to printer or PC via DataWay software
* With cell constant (K)=10
High specification laboratory conductivity/resistivity/TDS/salinity/°C meter that offers additional accuracy with 1, 2 or 3 point conductivity calibration across a wide measurement range. The dedicated 'pure water' mode ensures the optimum accuracy for this difficult application.
Bestellinformation: 4510 and 4520 supplied with glass conductivity probe 663-0005 with ATC (K=1), electrode holder and power supply.