Spektralphotometer, 6300
Lieferant: Cole-Parmer
630531 83054-07
JENW630531EA 2670 EUR
JENW630531 JENW83054-07
Spektralphotometer, 6300
Spektrophotometer und Photometer UV/Vis-Spektralphotometer
The 6300 is designed to provide a compact, high quality, easy to use measurement system.
  • Large sample compartment for a wide range of sampling devices to be fitted
  • Simple keypad layout and menu operating protocols
  • Display simultaneously shows both wavelength and the selected photometric value
  • Absorbance, transmittance and concentration measurement modes
Suitable for routine analysis in schools and colleges, as well as in QC and testing procedures for a wide variety of services and industries.
The 6320D model is based on the 6300 visible spectrophotometer platform and includes a domed lid which accepts taller tubes up to a height of 105 mm.
Lieferumfang: Model 6300 supplied with mains lead, 100 disposable cuvettes, 10×10 mm cell holder, interface cable and PC application software on CD-ROM.
Model 6320D units supplied with mains lead, dual cell holder (for 10 mm square cuvettes and 12,7 mm Ø tubes).