No-Met biocompatible mobile phase filters
Lieferant: VICI JOUR RESEARCH
JR-32171 JR-32172 JR-32179 JR-32178 JR-32174
JOURJR-32171EA 103 EUR
JOURJR-32171 JOURJR-32172 JOURJR-32179 JOURJR-32178 JOURJR-32174
No-Met biocompatible mobile phase filters
Filter Flüssigkeits-Prozessfilter
The No-Met polyethylene filter is designed for applications involving the separation of biomolecules, with inert polymeric fittings for 1/8" tubing and a 20 µm filter effectively eliminating metal contamination from the fluid path.
  • Inert and biocompatible
  • Recommended for IC and biochromatography applications
  • Can be used at flow rates up to 500 ml/min