Safety cap
Lieferant: VICI JOUR RESEARCH
JR-9000-0001
JOURJR-9000-0001EA 55.1 EUR
JOURJR-9000-0001
Safety cap
Verschlüsse Safety Caps und Safety Waste Caps
This cap can be used for helium sparging and for delivering HPLC mobile phases. The tubing ports are made so that you push the tubing through the hole, while 1/4-28 fittings provide the best connection. Unused ports can be plugged as required.