Waste caps
Lieferant: VICI JOUR RESEARCH
JR-S-15313 JR-S-15303 JR-S-15113 JR-S-15103 JR-S-16103 JR-S-16003 JR-S-16013 JR-S-18314 JR-S-18304 JR-S-18014 JR-S-18004 JR-S-16113 JR-S-11023
JOURJR-S-15313EA 318 EUR
JOURJR-S-15313 JOURJR-S-15303 JOURJR-S-15113 JOURJR-S-15103 JOURJR-S-16103 JOURJR-S-16003 JOURJR-S-16013 JOURJR-S-18314 JOURJR-S-18304 JOURJR-S-18014 JOURJR-S-18004 JOURJR-S-16113 JOURJR-S-11023
Waste caps
Verschlüsse Safety Caps und Safety Waste Caps
These waste caps include three or four ports for ¹/₈ and ¹/₁₆" tubing. Two versions of each waste cap are available, with an extra port to accommodate either a 1/2-20 port for ¹/₄" O.Ø tubing, or a barbed hose adapter.
Bestellinformation: Barbed adapters must be ordered separately.