Browguards with 20 cm polycarbonate visor, Invincible®
Lieferant: JSP
AFA061-131-100
JSPAAFA061-131-100EA 69.2 EUR
JSPAAFA061-131-100
Browguards with 20 cm polycarbonate visor, Invincible®
Gesichtsschutz
Faceshield offers protection against medium energy, high-speed particles at 120 m/s (270 mph), liquid droplets, and molten metal.
  • Polycarbonate browguard
  • Aluminium edged polycarbonate visor
  • Wheel ratchet headband
  • Optical class 1
A polycarbonate browguard with nylon wheel ratchet adjustable head harness and aluminium edged clear polycarbonate visor, offering protection against medium energy, high-speed particles at 120 m/s (270 mph), liquid droplets, and molten metal. The optical qualities of the visor comply with the requirements of optical class 1 (highest).
Standards: Conforms to EN 166.1.B.3.9.