Double lens goggle, Thermex™
Lieferant: JSP
AGM020-723-000
JSPAAGM020-723-000EA 52.3 EUR
JSPAAGM020-723-000
Double lens goggle, Thermex™
Vollsichtbrillen
The Thermex™ is an advanced double lensed goggle with TMC™ technology that offers fog-free use, even in extreme temperatures.
  • K&N Class lens
  • TMC™ technology provides fog-free use
  • Optical class 1
  • 30 mm elastic headband
  • PVC frame
  • Dust, liquid splash, molten metal, and extreme temperature protection
  • 99.9% protection against UV radiation
Thermex™ goggles offer protection against medium energy, high-speed particles at 120 m/s (270 mph), and protection against high-speed particles at extreme temperatures. The optical qualities of the lens comply with the requirements of optical class 1 (highest). K & N Class lens also offer anti-scratch and anti-mist protection. They also offers 99.9% protection against UV radiation.
NBR foam on the goggle housing provides a comfortable fitting to a wide range of facial profiles. Thermex™ goggles also feature a wide 30mm elastic headband for improved comfort and secure fit. Can be used with an RX insert for prescription eyewear.
Low profile design means the Thermex™ goggle can be worn with an industrial safety helmet or bumpcap, and is fully compatible with our Evolution® range of safety helmets and our Hardcap™ A1+ baseball style bumpcaps.
Standards: EN 166 1.B.T.3.4.9.K.N and EN 170.