Double lens goggle, EVO®
Lieferant: JSP
AGM020-823-000
JSPAAGM020-823-000EA 41.1 EUR
JSPAAGM020-823-000
Double lens goggle, EVO®
Vollsichtbrillen
Double lens goggle engineered to offer outstanding performance, compatibility, comfort, and looks.
  • Double lens design
  • Indirect venting
  • PVC frame
  • Elastic headband
  • Contour design
  • High level of protection
  • Fully compatible with the JSP EVO® Range of industrial safety helmets
Double lens design for optimum anti fog protection, as well as anti-mist and scratch resistant lens with UV protection. Low profile design is lightweight with soft EVA foam housing for exceptional wearer comfort. Googles provide dust, liquid splash, molten metal, and extreme temperature protection. Compatible with prescription lenses by using RX Inserts for EVO® Goggles.
Standards: EN 166; EN 170.