Safety helmet with CR2 reflective decals and ID badge holder, EVOLite®
Lieferant: JSP
AJB170-D00-100
JSPAAJB170-D00-100EA 72.1 EUR
JSPAAJB170-D00-100
Safety helmet with CR2 reflective decals and ID badge holder, EVOLite®
Schutzhelme Schutzhelme
EVOLite® is one of the lightest safety helmets for sale in the UK, conforming to EN 397. This model is fitted with CR2 reflective decals and ID badge holder.
  • Lightweight industrial safety helmet fitted with CR2 reflective decals
  • Tough ABS shell
  • Vented design
  • Integrated ID badge holder
  • Wheel ratchet adjustment
  • Chamlon sweatband
The EVOLite® safety helmet is lightweight, extremely comfortable, and fits the widest range of head shapes and sizes. The harness and shell are truly integrated as one to give the best performance by keeping it firmly rooted on the head, even when fully inverted. Fitted with CR2 reflective decals which offer on average 60% more reflectivity than standard reflective clothing. This model is also fitted with integrated ID badge holder, suitable for standard credit card sized ID's.
Weighing less than 300 g (dependent on model), the EVOLite® is the lightest safety helmet for sale in the UK. A 6-point terylene cradle harness system offers unrivalled comfort without compromising performance. Egyptian cotton core with porous PU coating for maximum sweat absorption. pH neutral, dermatologically tested. 3-D precision fitting uses the unique 1-2-3 point harness depth settings.
Standards: EN 397 and EN 12899-1 (reflective).