PAPR systems, PowerCap® Infinity®
Lieferant: JSP
CAU660-000-400 CEA646-000-100
JSPACAU660-000-400EA 94.6 EUR
JSPACAU660-000-400 JSPACEA646-000-100
PAPR systems, PowerCap® Infinity®
Atemschutzmasken Atemschutz-Systeme
A fully integrated TH3 powered air respirator offering four of the highest levels of protection in one intuitive head-mounted unit. The system provides complete above the neck 4-in-1 protection; respiratory, head, eye, and face.
  • 4-in-1 protection
  • Super strong ABS shell
  • Superior all-day protection
  • Optical class 1 visor
  • High capacity HEPA filters
  • Polycarbonate face shield
  • Rubber skirt
PowerCap® Infinity® is the perfect choice for many industries where dust poses a great danger to health. PowerCap® Infinity® removes the need for face fit testing,More Product Information